Assay Details
| Gene Symbol: CALCB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001149.2 | NP_001001149.1 | 1 - 2 | 111 | 102 | |
| NM_001134662.1 | NP_001128134.1 | 1 - 2 | 111 | 102 | ||
| GenBank mRNA | AB125101.1 | - | 1 - 2 | 98 | 102 | |
| BC118475.1 | - | 1 - 2 | 112 | 102 | ||
| DV827218.1 | - | 1 - 2 | 112 | 102 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC118475.1, NM_001001149.2, NM_001134662.1 transcripts.
Set Membership: |
|

CALCB
Genomic Map