Assay Details
| Gene Symbol: CALCB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001149.2 | NP_001001149.1 | 4 - 5 | 520 | 96 | |
| NM_001134662.1 | NP_001128134.1 | 4 - 5 | 551 | 96 | ||
| GenBank mRNA | AB125101.1 | - | 4 - 5 | 507 | 96 | |
| BC118475.1 | - | 4 - 5 | 552 | 96 | ||
| DV827218.1 | - | 4 - 5 | 552 | 96 | ||
More Information
Set Membership: |
|

CALCB
Genomic Map