Assay Details
| Gene Symbol: HSP90AA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012670.2 | NP_001012688.1 | 5 - 6 | 1123 | 79 | |
| GenBank mRNA | AB072368.1 | - | 5 - 6 | 1090 | 79 | |
More Information
Set Membership: |
|

HSP90AA1
Genomic Map