Assay Details
| Gene Symbol: BOLA-DRA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012677.1 | NP_001012695.1 | 1 - 2 | 159 | 72 | |
| GenBank mRNA | BC102053.1 | - | 1 - 2 | 143 | 72 | |
| BC142119.1 | - | 1 - 2 | 163 | 72 | ||
| BT021059.1 | - | 1 - 2 | 159 | 72 | ||
| D37955.1 | - | 1 - 2 | 32 | 72 | ||
| D37956.1 | - | 1 - 2 | 112 | 72 | ||
| X78308.1 | - | 90 | 72 | |||
More Information
Set Membership: |
|

BOLA-DRA
Genomic Map