Assay Details
| Gene Symbol: BOLA-DRA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012677.1 | NP_001012695.1 | 4 - 5 | 858 | 100 | |
| GenBank mRNA | BC102053.1 | - | 4 - 5 | 842 | 100 | |
| BC142119.1 | - | 4 - 5 | 862 | 100 | ||
| BT021059.1 | - | 4 - 5 | 858 | 100 | ||
| D37955.1 | - | 4 - 5 | 731 | 100 | ||
| D37956.1 | - | 4 - 5 | 811 | 100 | ||
| X78308.1 | - | 789 | 100 | |||
More Information
Set Membership: |
|

BOLA-DRA
Genomic Map