Assay Details
| Gene Symbol: CAPZA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012998.2 | NP_001013016.1 | 1 - 2 | 102 | 67 | |
| GenBank mRNA | BC102732.2 | - | 1 - 2 | 99 | 67 | |
| BT021023.1 | - | 1 - 2 | 102 | 67 | ||
More Information
Set Membership: |
|

CAPZA2
Genomic Map