Assay Details
| Gene Symbol: COMMD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014850.1 | NP_001014850.1 | 1 - 2 | 82 | 91 | |
| GenBank mRNA | BT020919.1 | - | 1 - 2 | 82 | 91 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001014850.1 transcripts.
Set Membership: |
|

COMMD5
Genomic Map