Assay Details
| Gene Symbol: RNF34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014858.1 | NP_001014858.1 | 2 - 3 | 309 | 94 | |
| GenBank mRNA | BC110246.1 | - | 3 - 4 | 563 | 94 | |
| BT020924.1 | - | 2 - 3 | 309 | 94 | ||
More Information
Set Membership: |
|

RNF34
Genomic Map