Assay Details
| Gene Symbol: CDIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014892.1 | NP_001014892.1 | 4 - 5 | 526 | 74 | |
| GenBank mRNA | BC102294.1 | - | 4 - 5 | 439 | 74 | |
| BT021752.1 | - | 4 - 5 | 526 | 74 | ||
More Information
Set Membership: |
|

CDIP1
Genomic Map