Assay Details
| Gene Symbol: TXNL4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014897.2 | NP_001014897.1 | 3 - 4 | 541 | 85 | |
| GenBank mRNA | BC103293.1 | - | 3 - 4 | 541 | 85 | |
| BT020765.1 | - | 3 - 4 | 541 | 85 | ||
More Information
Set Membership: |
|

TXNL4B
Genomic Map