Assay Details
| Gene Symbol: BYSL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014924.1 | NP_001014924.1 | 1 - 2 | 539 | 60 | |
| GenBank mRNA | BC151370.1 | - | 1 - 2 | 592 | 60 | |
| BT020890.1 | - | 1 - 2 | 539 | 60 | ||
More Information
Set Membership: |
|

BYSL
Genomic Map