Assay Details
| Gene Symbol: RPL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014928.1 | NP_001014928.1 | 6 - 7 | 750 | 145 | |
| GenBank mRNA | BT021516.1 | - | 6 - 7 | 750 | 145 | |
More Information
Set Membership: |
|

RPL7
Genomic Map