Assay Details
| Gene Symbol: ORMDL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014931.1 | NP_001014931.1 | 1 - 2 | 64 | 119 | |
| GenBank mRNA | BC109992.1 | - | 1 - 2 | 73 | 119 | |
| BT021048.1 | - | 1 - 2 | 64 | 119 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC109992.1, NM_001014931.1 transcripts.
Set Membership: |
|

ORMDL2
Genomic Map