Assay Details
| Gene Symbol: ORMDL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014931.1 | NP_001014931.1 | 2 - 3 | 235 | 78 | |
| GenBank mRNA | BC109992.1 | - | 2 - 3 | 244 | 78 | |
| BT021048.1 | - | 2 - 3 | 235 | 78 | ||
More Information
Set Membership: |
|

ORMDL2
Genomic Map