Assay Details
| Gene Symbol: RARA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014942.4 | NP_001014942.2 | 4 - 5 | 990 | 67 | |
| GenBank mRNA | BT021634.1 | - | 4 - 5 | 839 | 67 | |
More Information
Set Membership: |
|

RARA
Genomic Map