Assay Details
| Gene Symbol: C1QA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014945.2 | NP_001014945.1 | 2 - 3 | 202 | 65 | |
| GenBank mRNA | AY911382.1 | - | 2 - 3 | 203 | 65 | |
| BC105345.1 | - | 2 - 3 | 203 | 65 | ||
| BT020977.1 | - | 2 - 3 | 246 | 65 | ||
More Information
Set Membership: |
|

C1QA
Genomic Map