Assay Details
| Gene Symbol: ID3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014950.1 | NP_001014950.1 | 1 - 2 | 345 | 128 | |
| GenBank mRNA | BC102157.1 | - | 1 - 2 | 369 | 128 | |
| BT021039.1 | - | 1 - 2 | 345 | 128 | ||
More Information
Set Membership: |
|

ID3
Genomic Map