Assay Details
| Gene Symbol: ARAF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014964.1 | NP_001014964.1 | 14 - 15 | 1649 | 58 | |
| GenBank mRNA | BC105445.1 | - | 14 - 15 | 1671 | 58 | |
| BT021200.1 | - | 14 - 15 | 1649 | 58 | ||
More Information
Set Membership: |
|

ARAF
Genomic Map