Assay Details
| Gene Symbol: NDN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014982.1 | NP_001014982.1 | 1 | 1446 | 91 | |
| GenBank mRNA | BC146188.1 | - | 1 | 1461 | 91 | |
| BT020838.1 | - | 1 | 1446 | 91 | ||
| BT029863.1 | - | 1 | 1466 | 91 | ||
More Information
Set Membership: |
|

NDN
Genomic Map