Assay Details
| Gene Symbol: CHID1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015515.1 | NP_001015515.1 | 10 - 11 | 1045 | 54 | |
| GenBank mRNA | BC118449.1 | - | 10 - 11 | 1055 | 54 | |
| BT020630.1 | - | 10 - 11 | 1031 | 54 | ||
| BT020655.1 | - | 10 - 11 | 1045 | 54 | ||
| BT020789.1 | - | 9 - 10 | 1016 | 54 | ||
| BT020873.1 | - | 10 - 11 | 1045 | 54 | ||
| BT021170.1 | - | 10 - 11 | 1045 | 54 | ||
| BT021178.1 | - | 9 - 10 | 1011 | 54 | ||
| BT021793.1 | - | 10 - 11 | 1044 | 54 | ||
More Information
Set Membership: |
|

CHID1
Genomic Map