Assay Details
| Gene Symbol: ABRAXAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015516.1 | NP_001015516.1 | 1 - 2 | 95 | 108 | |
| GenBank mRNA | BC123849.1 | - | 1 - 2 | 110 | 108 | |
| BT020879.1 | - | 1 - 2 | 95 | 108 | ||
More Information
Set Membership: |
|

ABRAXAS1
Genomic Map