Assay Details
| Gene Symbol: PRR5L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015519.1 | NP_001015519.1 | 4 - 5 | 558 | 77 | |
| GenBank mRNA | BC151482.1 | - | 4 - 5 | 549 | 77 | |
| BT020860.1 | - | 4 - 5 | 558 | 77 | ||
More Information
Set Membership: |
|

PRR5L
Genomic Map