Assay Details
| Gene Symbol: MVK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015528.1 | NP_001015528.1 | 2 - 3 | 160 | 72 | |
| GenBank mRNA | BC104540.1 | - | 2 - 3 | 161 | 72 | |
| BT020667.1 | - | 2 - 3 | 160 | 72 | ||
| BT020832.1 | - | 2 - 3 | 160 | 72 | ||
| BT021860.1 | - | 2 - 3 | 162 | 72 | ||
More Information
Set Membership: |
|

MVK
Genomic Map