Assay Details
| Gene Symbol: NPRL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015547.1 | NP_001015547.1 | 1 - 2 | 157 | 117 | |
| GenBank mRNA | BC105243.1 | - | 1 - 2 | 201 | 117 | |
| BT020747.1 | - | 1 - 2 | 157 | 117 | ||
| BT020810.1 | - | 1 - 2 | 152 | 117 | ||
| BT020821.1 | - | 1 - 2 | 130 | 117 | ||
More Information
Set Membership: |
|

NPRL2
Genomic Map