Assay Details
| Gene Symbol: AADAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015551.1 | NP_001015551.1 | 8 - 9 | 1064 | 66 | |
| GenBank mRNA | BC109974.1 | - | 8 - 9 | 1122 | 66 | |
| BT020844.1 | - | 8 - 9 | 1064 | 66 | ||
| BT020886.1 | - | 8 - 9 | 1032 | 66 | ||
More Information
Set Membership: |
|

AADAT
Genomic Map