Assay Details
| Gene Symbol: OSER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015568.1 | NP_001015568.1 | 2 - 3 | 191 | 82 | |
| GenBank mRNA | BC110226.1 | - | 2 - 3 | 252 | 82 | |
| BT021020.1 | - | 2 - 3 | 191 | 82 | ||
More Information
Set Membership: |
|

OSER1
Genomic Map