Assay Details
| Gene Symbol: PARK7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015572.1 | NP_001015572.1 | 6 - 7 | 519 | 75 | |
| GenBank mRNA | BC102707.1 | - | 6 - 7 | 540 | 75 | |
| BT021074.1 | - | 6 - 7 | 519 | 75 | ||
More Information
Set Membership: |
|

PARK7
Genomic Map