Assay Details
| Gene Symbol: SFXN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015574.1 | NP_001015574.1 | 10 - 11 | 949 | 59 | |
| GenBank mRNA | BC103181.1 | - | 10 - 11 | 963 | 59 | |
| BT020892.1 | - | 10 - 11 | 949 | 59 | ||
| KX258203.1 | - | 10 - 11 | 949 | 59 | ||
More Information
Set Membership: |
|

SFXN1
Genomic Map