Assay Details
| Gene Symbol: APEX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015577.1 | NP_001015577.1 | 5 - 6 | 651 | 61 | |
| GenBank mRNA | BT020881.1 | - | 5 - 6 | 651 | 61 | |
| BT021707.1 | - | 5 - 6 | 642 | 61 | ||
More Information
Set Membership: |
|

APEX2
Genomic Map