Assay Details
| Gene Symbol: B4GALT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015609.1 | NP_001015609.1 | 3 - 4 | 739 | 65 | |
| GenBank mRNA | BC123718.1 | - | 4 - 5 | 710 | 65 | |
| BT020682.1 | - | 4 - 5 | 717 | 65 | ||
| BT020868.1 | - | 3 - 4 | 739 | 65 | ||
| BT020916.1 | - | 4 - 5 | 711 | 65 | ||
| BT026158.1 | - | 4 - 5 | 711 | 65 | ||
More Information
Set Membership: |
|

B4GALT3
Genomic Map