Assay Details
| Gene Symbol: GABRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015618.1 | NP_001015618.1 | 9 - 10 | 1180 | 69 | |
| GenBank mRNA | BC123810.1 | - | 9 - 10 | 1183 | 69 | |
| BT020763.1 | - | 9 - 10 | 1180 | 69 | ||
More Information
Set Membership: |
|

GABRP
Genomic Map