Assay Details
| Gene Symbol: DHODH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015650.1 | NP_001015650.1 | 2 - 3 | 238 | 68 | |
| GenBank mRNA | BC120337.1 | - | 2 - 3 | 238 | 68 | |
| BT020807.1 | - | 2 - 3 | 238 | 68 | ||
| BT026309.1 | - | 2 - 3 | 227 | 68 | ||
More Information
Set Membership: |
|

DHODH
Genomic Map