Assay Details
| Gene Symbol: MGAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015653.1 | NP_001015653.1 | 1 - 2 | 80 | 58 | |
| GenBank mRNA | BT020936.1 | - | 1 - 2 | 80 | 58 | |
| BT021819.1 | - | 1 - 2 | 447 | 58 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001015653.1 transcripts.
Set Membership: |
|

MGAT1
Genomic Map