Assay Details
| Gene Symbol: C3H1orf43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015666.1 | NP_001015666.1 | 2 - 3 | 318 | 134 | |
| GenBank mRNA | BC118321.1 | - | 2 - 3 | 305 | 134 | |
| BT021077.1 | - | 2 - 3 | 318 | 134 | ||
More Information
Set Membership: |
|

C3H1orf43
Genomic Map