Assay Details
| Gene Symbol: CDIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015668.1 | NP_001015668.1 | 2 - 3 | 288 | 71 | |
| GenBank mRNA | BC142022.1 | - | 2 - 3 | 368 | 71 | |
| BT020842.1 | - | 2 - 3 | 288 | 71 | ||
More Information
Set Membership: |
|

CDIN1
Genomic Map