Assay Details
| Gene Symbol: CDIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015668.1 | NP_001015668.1 | 6 - 7 | 572 | 71 | |
| GenBank mRNA | BC134779.1 | - | 3 - 4 | 178 | 71 | |
| BC142022.1 | - | 6 - 7 | 652 | 71 | ||
| BT020842.1 | - | 6 - 7 | 572 | 71 | ||
More Information
Set Membership: |
|

CDIN1
Genomic Map