Assay Details
| Gene Symbol: AP5M1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015670.1 | NP_001015670.1 | 3 - 4 | 1193 | 77 | |
| GenBank mRNA | BT020795.1 | - | 3 - 4 | 1193 | 77 | |
More Information
Set Membership: |
|

AP5M1
Genomic Map