Assay Details
| Gene Symbol: MYOZ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015676.1 | NP_001015676.1 | 4 - 5 | 512 | 101 | |
| GenBank mRNA | BC103121.1 | - | 4 - 5 | 513 | 101 | |
| BT020817.1 | - | 4 - 5 | 512 | 101 | ||
More Information
Set Membership: |
|

MYOZ2
Genomic Map