Assay Details
| Gene Symbol: NAPEPLD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015680.1 | NP_001015680.1 | 2 - 3 | 433 | 90 | |
| GenBank mRNA | BT021908.1 | - | 2 - 3 | 433 | 90 | |
More Information
Set Membership: |
|

NAPEPLD
Genomic Map