Assay Details
| Gene Symbol: GNPTG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017428.1 | NP_001017428.1 | 2 - 3 | 124 | 84 | |
| GenBank mRNA | BC142266.1 | - | 3 - 4 | 203 | 84 | |
More Information
Set Membership: |
|

GNPTG
Genomic Map