Assay Details
| Gene Symbol: RAB25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017936.1 | NP_001017936.1 | 2 - 3 | 466 | 80 | |
| GenBank mRNA | BC120091.1 | - | 2 - 3 | 466 | 80 | |
| BT021481.1 | - | 2 - 3 | 466 | 80 | ||
More Information
Set Membership: |
|

RAB25
Genomic Map