Assay Details
| Gene Symbol: HYAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017941.1 | NP_001017941.1 | 2 - 3 | 1030 | 79 | |
| GenBank mRNA | BC102473.1 | - | 2 - 3 | 1094 | 79 | |
| BT020775.1 | - | 2 - 3 | 1029 | 79 | ||
| BT021035.1 | - | 2 - 3 | 1030 | 79 | ||
More Information
Set Membership: |
|

HYAL1
Genomic Map