Assay Details
| Gene Symbol: B4GALNT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017943.1 | NP_001017943.1 | 10 - 11 | 1768 | 85 | |
| GenBank mRNA | BC142194.1 | - | 10 - 11 | 1775 | 85 | |
| BT021615.1 | - | 10 - 11 | 1768 | 85 | ||
More Information
Set Membership: |
|

B4GALNT1
Genomic Map