Assay Details
| Gene Symbol: LSM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017952.1 | NP_001017952.1 | 2 - 3 | 285 | 97 | |
| GenBank mRNA | BC110194.1 | - | 2 - 3 | 281 | 97 | |
| BT020738.1 | - | 2 - 3 | 205 | 97 | ||
| BT020771.1 | - | 2 - 3 | 293 | 97 | ||
| BT020779.1 | - | 2 - 3 | 285 | 97 | ||
More Information
Set Membership: |
|

LSM1
Genomic Map