Assay Details
| Gene Symbol: RPL27A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024471.2 | NP_001019642.1 | 4 - 5 | 335 | 88 | |
| GenBank mRNA | AY911324.1 | - | 4 - 5 | 336 | 88 | |
| BC109683.1 | - | 1 | 337 | 88 | ||
| BT021568.1 | - | 3 - 4 | 717 | 88 | ||
| X97045.1 | - | 334 | 88 | |||
More Information
Set Membership: |
|

RPL27A
Genomic Map