Assay Details
| Gene Symbol: IQCC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024492.1 | NP_001019663.1 | 3 - 4 | 1093 | 97 | |
| GenBank mRNA | BC110130.1 | - | 4 - 5 | 582 | 97 | |
| BT021671.1 | - | 3 - 4 | 1093 | 97 | ||
More Information
Set Membership: |
|

IQCC
Genomic Map