Assay Details
| Gene Symbol: NECTIN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024494.1 | NP_001019665.1 | 2 - 3 | 689 | 97 | |
| GenBank mRNA | BC148055.1 | - | 2 - 3 | 686 | 97 | |
| BT020770.1 | - | 2 - 3 | 689 | 97 | ||
More Information
Set Membership: |
|

NECTIN4
Genomic Map