Assay Details
| Gene Symbol: MRPS17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024500.2 | NP_001019671.1 | 2 - 3 | 237 | 118 | |
| GenBank mRNA | AY911326.1 | - | 2 - 3 | 238 | 118 | |
| BC111630.1 | - | 2 - 3 | 186 | 118 | ||
More Information
Set Membership: |
|

MRPS17
Genomic Map