Assay Details
| Gene Symbol: ZGPAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024514.2 | NP_001019685.2 | 2 - 3 | 720 | 59 | |
| GenBank mRNA | BC118131.1 | - | 2 - 3 | 712 | 59 | |
| BT021717.1 | - | 1 - 2 | 801 | 59 | ||
| EH162907.1 | - | 2 - 3 | 719 | 59 | ||
More Information
Set Membership: |
|

ZGPAT
Genomic Map