Assay Details
| Gene Symbol: ALKBH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024516.1 | NP_001019687.1 | 2 - 3 | 493 | 88 | |
| GenBank mRNA | BC120178.1 | - | 2 - 3 | 524 | 88 | |
| BT021573.1 | - | 2 - 3 | 493 | 88 | ||
More Information
Set Membership: |
|

ALKBH2
Genomic Map